Assay Details
| Gene Symbol: Tsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289575.1 | NP_001276504.1 | 17 - 18 | 2424 | 84 | |
| NM_001289576.1 | NP_001276505.1 | 17 - 18 | 2406 | 84 | ||
| NM_022887.4 | NP_075025.2 | 17 - 18 | 2392 | 84 | ||
| GenBank mRNA | AB047561.1 | - | 15 - 16 | 2191 | 84 | |
| AJ271912.1 | - | 17 - 18 | 2416 | 84 | ||
| AK030318.1 | - | 17 - 18 | 2389 | 84 | ||
| AK122229.1 | - | 3 - 4 | 716 | 84 | ||
| AK147428.1 | - | 17 - 18 | 2397 | 84 | ||
| BC052399.1 | - | 16 - 17 | 2345 | 5720967 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsc1
Genomic Map