Assay Details
| Gene Symbol: Tsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289575.1 | NP_001276504.1 | 18 - 19 | 2614 | 89 | |
| NM_001289576.1 | NP_001276505.1 | 18 - 19 | 2596 | 89 | ||
| NM_022887.4 | NP_075025.2 | 18 - 19 | 2582 | 89 | ||
| GenBank mRNA | AB047561.1 | - | 16 - 17 | 2381 | 89 | |
| AJ271912.1 | - | 18 - 19 | 2606 | 89 | ||
| AK030318.1 | - | 18 - 19 | 2579 | 89 | ||
| AK122229.1 | - | 4 - 5 | 906 | 89 | ||
| AK147428.1 | - | 18 - 19 | 2587 | 89 | ||
| BC052399.1 | - | 17 - 18 | 2535 | 5720967 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsc1
Genomic Map