Assay Details
| Gene Symbol: Gbgt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139197.2 | NP_631936.2 | 4 - 5 | 457 | 103 | |
| GenBank mRNA | AB084789.1 | - | 3 - 4 | 206 | 103 | |
| AF292399.1 | - | 3 - 4 | 186 | 103 | ||
| BC106831.2 | - | 3 - 4 | 186 | 40044790 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gbgt1
Genomic Map