Assay Details
| Gene Symbol: Sfmbt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198808.1 | NP_001185737.1 | 10 - 11 | 1312 | 97 | |
| NM_177386.5 | NP_796360.2 | 11 - 12 | 1435 | 97 | ||
| GenBank mRNA | AK052339.1 | - | 10 - 11 | 1379 | 98 | |
| AK147557.1 | - | 10 - 11 | 1317 | 97 | ||
| AK147651.1 | - | 11 - 12 | 1290 | 97 | ||
| AK220408.1 | - | 8 - 9 | 1177 | 97 | ||
| BC139354.1 | - | 10 - 11 | 1223 | 8862376 | 97 | |
| BC145610.1 | - | 9 - 10 | 1179 | 9054176 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sfmbt2
Genomic Map