Assay Details
| Gene Symbol: Ndufaf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027093.4 | NP_081369.2 | 7 - 8 | 778 | 122 | |
| GenBank mRNA | AK009123.1 | - | 7 - 8 | 845 | 122 | |
| BC005630.1 | - | 7 - 8 | 645 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndufaf5
Genomic Map