Assay Details
| Gene Symbol: Nsun6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165941.1 | NP_001159413.1 | 9 - 10 | 1357 | 94 | |
| NM_001165943.1 | NP_001159415.1 | 8 - 9 | 1061 | 94 | ||
| NM_028950.4 | NP_083226.3 | 8 - 9 | 1244 | 94 | ||
| GenBank mRNA | AK016629.1 | - | 8 - 9 | 1061 | 94 | |
| AK016811.1 | - | 9 - 10 | 1370 | 94 | ||
| AK077778.1 | - | 8 - 9 | 1128 | 94 | ||
| AK132428.1 | - | 8 - 9 | 1211 | 94 | ||
| BC025622.1 | - | 5 - 6 | 587 | 94 | ||
| BC053751.1 | - | 8 - 9 | 1212 | 6335896 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nsun6
Genomic Map