Assay Details
| Gene Symbol: Wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144783.2 | NP_659032.3 | 7 - 8 | 1522 | 61 | |
| GenBank mRNA | BG087452.2 | - | 3 - 4 | 321 | 61 | |
| DQ537939.1 | - | 7 - 8 | 859 | 61 | ||
| M55512.1 | - | 7 - 8 | 1527 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wt1
Genomic Map