Assay Details
| Gene Symbol: Foxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI448353.1 | - | 1 - 2 | 39 | 74 | |
| AK053772.1 | - | 1 - 2 | 114 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 26839 |
Gm26839 | Anti-sense | Chr. 12 - 99560295 - 99620033 on NCBI Build GRCm38 | ||
| Downstream | RIKEN cDNA 1700064M15 gene | 1700064M15Rik | 62.5kb | Sense | Chr. 12 - 99626053 - 99627974 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Foxn3
Genomic Map