Assay Details
| Gene Symbol: Ptpn21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146199.1 | NP_001139671.1 | 16 - 17 | 3235 | 72 | |
| NM_011877.2 | NP_036007.2 | 16 - 17 | 3370 | 72 | ||
| GenBank mRNA | BC049893.1 | - | 14 - 15 | 3117 | 72 | |
| BC055942.1 | - | 4 - 5 | 1759 | 72 | ||
| BC066856.1 | - | 14 - 15 | 1629 | 72 | ||
| BC098500.1 | - | 16 - 17 | 3235 | 30081068 | 72 | |
| D37801.1 | - | 16 - 17 | 3319 | 72 | ||
| D83072.1 | - | 3 - 4 | 631 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn21
Genomic Map