Assay Details
| Gene Symbol: Cntrob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172560.3 | NP_766148.2 | 10 - 11 | 2346 | 89 | |
| GenBank mRNA | AK036710.1 | - | 9 - 10 | 2166 | 89 | |
| AK147774.1 | - | 10 - 11 | 2232 | 89 | ||
| BC058512.1 | - | 10 - 11 | 1179 | 6817014 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cntrob
Genomic Map