Assay Details
| Gene Symbol: Cntrob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172560.3 | NP_766148.2 | 7 - 8 | 1817 | 65 | |
| GenBank mRNA | AK036710.1 | - | 6 - 7 | 1637 | 65 | |
| AK147774.1 | - | 7 - 8 | 1703 | 65 | ||
| BC058512.1 | - | 7 - 8 | 650 | 6817014 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cntrob
Genomic Map