| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100007677.mm.1 |
NM_001024206.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007677.mm.1 |
NM_001024206.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007677.mm.1 |
TC1100000907.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100007676.mm.1 |
NM_001024206.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007676.mm.1 |
NM_001024206.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007676.mm.1 |
TC1100000907.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100007678.mm.1 |
NM_001024206.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007678.mm.1 |
NM_001024206.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007678.mm.1 |
TC1100000907.mm.1 |
MTA TC |