Assay Details
| Gene Symbol: Serpina3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173024.3 | NP_766612.1 | 1 - 2 | 79 | 69 | |
| GenBank mRNA | AK037160.1 | - | 1 - 2 | 81 | 69 | |
| BC125418.1 | - | 1 - 2 | 62 | 40129933 | 69 | |
| BC145808.1 | - | 1 - 2 | 62 | 40131111 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037160.1, BC125418.1, BC145808.1, NM_173024.3 transcripts.
Set Membership: |
|

Serpina3b
Genomic Map