Assay Details
| Gene Symbol: Ttyh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB162929.1 | - | 8 - 9 | 946 | 74 | |
| AF190699.1 | - | 8 - 9 | 1018 | 74 | ||
| AF190991.1 | - | 8 - 9 | 1018 | 74 | ||
| AK015486.1 | - | 7 - 8 | 669 | 74 | ||
| AK018148.1 | - | 8 - 9 | 1010 | 74 | ||
| AK031550.1 | - | 8 - 9 | 1046 | 74 | ||
| AK042758.1 | - | 8 - 9 | 799 | 74 | ||
| AK043998.1 | - | 8 - 9 | 1066 | 74 | ||
| AK048413.1 | - | 8 - 9 | 1017 | 74 | ||
| AK049903.1 | - | 7 - 8 | 752 | 74 | ||
| AK081981.1 | - | 8 - 9 | 1020 | 74 | ||
| AK083126.1 | - | 7 - 8 | 1143 | 74 | ||
| BC046310.1 | - | 8 - 9 | 1004 | 6307420 | 74 | |
| BC065694.1 | - | 8 - 9 | 1010 | 6813365 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttyh1
Genomic Map