Assay Details
| Gene Symbol: Eps8l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290416.1 | NP_001277345.1 | 10 - 11 | 1144 | 106 | |
| NM_026146.4 | NP_080422.1 | 10 - 11 | 1050 | 106 | ||
| GenBank mRNA | AK014556.1 | - | 10 - 11 | 1145 | 106 | |
| AK019490.2 | - | 10 - 11 | 1044 | 106 | ||
| AY074931.1 | - | 9 - 10 | 987 | 106 | ||
| BC027043.1 | - | 10 - 11 | 1334 | 106 | ||
| BC100383.1 | - | 10 - 11 | 1232 | 106 | ||
More Information
Set Membership: |
|

Eps8l1
Genomic Map