Assay Details
| Gene Symbol: Eps8l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290416.1 | NP_001277345.1 | 13 - 14 | 1511 | 124 | |
| NM_026146.4 | NP_080422.1 | 13 - 14 | 1417 | 124 | ||
| GenBank mRNA | AK014556.1 | - | 13 - 14 | 1512 | 124 | |
| AK019490.2 | - | 13 - 14 | 1411 | 124 | ||
| AY074931.1 | - | 12 - 13 | 1354 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eps8l1
Genomic Map