Assay Details
| Gene Symbol: Ppp1r12c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029834.3 | NP_084110.2 | 11 - 12 | 1547 | 79 | |
| GenBank mRNA | AK144691.1 | - | 11 - 12 | 1548 | 79 | |
| BC114396.1 | - | 1 - 2 | 64 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r12c
Genomic Map