Assay Details
| Gene Symbol: Ppp1r12c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029834.3 | NP_084110.2 | 12 - 13 | 1672 | 98 | |
| GenBank mRNA | AK046570.1 | - | 12 - 13 | 1643 | 98 | |
| AK144691.1 | - | 12 - 13 | 1673 | 98 | ||
| BC114396.1 | - | 2 - 3 | 189 | 98 | ||
More Information
Set Membership: |
|

Ppp1r12c
Genomic Map