Assay Details
| Gene Symbol: Zim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011769.4 | NP_035899.4 | 2 - 3 | 182 | 88 | |
| GenBank mRNA | AK086590.1 | - | 2 - 3 | 201 | 88 | |
| BC066196.1 | - | 2 - 3 | 182 | 30430385 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK086590.1, BC066196.1, NM_011769.4 transcripts.
Set Membership: |
|

Zim1
Genomic Map