| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700000766.mm.1 |
TC0700000070.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000766.mm.1 |
NM_001146024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000766.mm.1 |
NM_001146024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000760.mm.1 |
TC0700000070.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000760.mm.1 |
NM_001146024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000760.mm.1 |
NM_001146024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000760.mm.1 |
NM_028316.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000760.mm.1 |
NM_028316.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000765.mm.1 |
TC0700000070.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000765.mm.1 |
NM_001146024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000765.mm.1 |
NM_001146024.1 |
Refseq |