Assay Details
| Gene Symbol: Zfp444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146024.1 | NP_001139496.1 | 2 - 3 | 185 | 58 | |
| GenBank mRNA | AK012846.1 | - | 1 - 2 | 135 | 58 | |
| AK018079.1 | - | 2 - 3 | 187 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012846.1, NM_001146024.1 transcripts.
Set Membership: |
|

Zfp444
Genomic Map