Assay Details
| Gene Symbol: Tbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289689.1 | NP_001276618.1 | 5 - 6 | 929 | 114 | |
| GenBank mRNA | BC100305.1 | - | 5 - 6 | 886 | 30931957 | 114 |
| BC144904.1 | - | 5 - 6 | 907 | 9053452 | 114 | |
More Information
Set Membership: |
|

Tbpl2
Genomic Map