Assay Details
| Gene Symbol: Taf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027748.3 | NP_082024.2 | 2 - 3 | 622 | 105 | |
| GenBank mRNA | AJ292189.1 | - | 2 - 3 | 413 | 105 | |
| AK033848.1 | - | 2 - 3 | 272 | 105 | ||
| AK154372.1 | - | 2 - 3 | 631 | 105 | ||
| BC039989.1 | - | 2 - 3 | 578 | 105 | ||
| BC089030.1 | - | 2 - 3 | 461 | 105 | ||
| BC094546.1 | - | 2 - 3 | 579 | 105 | ||
| BC137617.1 | - | 2 - 3 | 563 | 8860636 | 105 | |
| BC137618.1 | - | 2 - 3 | 563 | 8860637 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taf3
Genomic Map