Assay Details
| Gene Symbol: Taf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027748.3 | NP_082024.2 | 5 - 6 | 2782 | 65 | |
| GenBank mRNA | AJ292189.1 | - | 5 - 6 | 2573 | 65 | |
| AK030878.1 | - | 3 - 4 | 1850 | 65 | ||
| AK084574.1 | - | 3 - 4 | 935 | 65 | ||
| AK136045.1 | - | 3 - 4 | 1440 | 65 | ||
| AK154372.1 | - | 5 - 6 | 2791 | 65 | ||
| BC089030.1 | - | 5 - 6 | 2621 | 65 | ||
| BC137617.1 | - | 5 - 6 | 2723 | 8860636 | 65 | |
| BC137618.1 | - | 5 - 6 | 2723 | 8860637 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taf3
Genomic Map