Assay Details
| Gene Symbol: 4930412O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024257.1 | 2 - 3 | 607 | 50 | ||
| GenBank mRNA | AK015124.1 | - | 2 - 3 | 602 | 50 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 13219 | Gm13219 | OTTMUSG00000011007 | -414.3kb | Sense | Chr. 2 - 9352413 - 9467458 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 9230102O04 gene | 9230102O04Rik | 177.0 | Anti-sense | Chr. 2 - 9883041 - 9889540 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015124.1 transcripts.
Set Membership: |
|

4930412O13Rik
Genomic Map