Assay Details
| Gene Symbol: H2afy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159514.1 | NP_001152986.1 | 4 - 5 | 716 | 70 | |
| NM_012015.2 | NP_036145.1 | 4 - 5 | 716 | 70 | ||
| GenBank mRNA | AB071988.2 | - | 4 - 5 | 652 | 70 | |
| AF171080.1 | - | 4 - 5 | 694 | 70 | ||
| BC006955.1 | - | 4 - 5 | 660 | 2811919 | 70 | |
More Information
Set Membership: |
|

H2afy
Genomic Map