Assay Details
| Gene Symbol: Tfap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122948.2 | NP_001116420.1 | 3 - 4 | 908 | 75 | |
| NM_001301674.1 | NP_001288603.1 | 4 - 5 | 887 | 75 | ||
| NM_011547.4 | NP_035677.2 | 3 - 4 | 643 | 75 | ||
| GenBank mRNA | AB221578.1 | - | 3 - 4 | 534 | 75 | |
| AK013900.1 | - | 1 - 2 | 64 | 75 | ||
| AK053704.1 | - | 3 - 4 | 893 | 75 | ||
| AK145772.1 | - | 3 - 4 | 598 | 75 | ||
| BC007471.1 | - | 3 - 4 | 618 | 3497082 | 75 | |
| BC018226.1 | - | 4 - 5 | 886 | 3983850 | 75 | |
| U17285.1 | - | 3 - 4 | 635 | 75 | ||
| U17286.1 | - | 2 - 3 | 114 | 75 | ||
| U17287.1 | - | 3 - 4 | 872 | 75 | ||
| U17288.1 | - | 2 - 3 | 657 | 75 | ||
| X74216.1 | - | 3 - 4 | 604 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tfap2a
Genomic Map