Assay Details
| Gene Symbol: Secisbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177608.3 | NP_808276.2 | 14 - 15 | 2270 | 88 | |
| GenBank mRNA | AK145135.1 | - | 14 - 15 | 2156 | 88 | |
| AK172920.1 | - | 14 - 15 | 2137 | 88 | ||
| BC071239.1 | - | 7 - 8 | 898 | 88 | ||
| BC141056.1 | - | 14 - 15 | 2230 | 9055668 | 88 | |
| CB528069.1 | - | 4 - 5 | 500 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Secisbp2l
Genomic Map