Assay Details
| Gene Symbol: Secisbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177608.3 | NP_808276.2 | 16 - 17 | 2649 | 87 | |
| GenBank mRNA | AK145135.1 | - | 16 - 17 | 2535 | 87 | |
| AK172920.1 | - | 16 - 17 | 2516 | 87 | ||
| BC141056.1 | - | 16 - 17 | 2609 | 9055668 | 87 | |
| CB289930.1 | - | 3 - 4 | 409 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Secisbp2l
Genomic Map