Assay Details
| Gene Symbol: 4930447K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046184.1 | 2 - 3 | 190 | 138 | ||
| GenBank mRNA | AK015408.1 | - | 2 - 3 | 190 | 138 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
tubulin, beta 2A class IIA |
Tubb2a |
M(beta)2, Tubb2 |
-17.2kb | Sense | Chr. 13 - 34074278 - 34078008 on NCBI Build GRCm38 |
| Downstream |
tubulin, beta 2B class IIB |
Tubb2b |
2410129E14Rik, brdp |
21.8kb | Sense | Chr. 13 - 34127008 - 34130354 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930447K03Rik
Genomic Map