Assay Details
| Gene Symbol: Crb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163566.1 | NP_001157038.1 | 6 - 7 | 1155 | 73 | |
| GenBank mRNA | BC043114.1 | - | 5 - 6 | 843 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crb2
Genomic Map