Assay Details
| Gene Symbol: Rmdn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033136.3 | NP_001028308.1 | 11 - 12 | 1419 | 84 | |
| GenBank mRNA | AK004786.1 | - | 11 - 12 | 1345 | 84 | |
| AK146299.1 | - | 11 - 12 | 1338 | 84 | ||
| AK151090.1 | - | 11 - 12 | 1340 | 84 | ||
| AK153322.1 | - | 11 - 12 | 1340 | 84 | ||
| AK153369.1 | - | 11 - 12 | 1339 | 84 | ||
| BC055754.1 | - | 11 - 12 | 1313 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233027 | BC055754.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233027 | AK151090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233027 | AK153322.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233027 | AK153369.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233027 | NM_001033136.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233027 | BC055754.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233027 | AK151090.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233027 | AK153322.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233027 | AK153369.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233027 | NM_001033136.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971804 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731262 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17489 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735371 | AK010581.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735371 | AK145843.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735371 | AK168384.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735371 | AK178869.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735371 | AK183750.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735371 | BC086644.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735371 | BC131945.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735371 | BC131973.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735371 | D82983.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735371 | NM_008820.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735371 | U51014.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103038 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371779 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816585 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 122090 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807601 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760307 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198838 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109030 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22567 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110136 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144128 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233027 | BC055754.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233027 | AK151090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233027 | AK153322.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233027 | AK153369.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233027 | NM_001033136.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233027 | BC055754.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233027 | AK151090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233027 | AK153322.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233027 | AK153369.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233027 | NM_001033136.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568165 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110136 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04664392 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144128 | - | - |
| Whole Mouse Genome G4122A | A_52_P333806 | BC055754.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P333806 | AK151090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P333806 | AK153322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P333806 | AK153369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P333806 | NM_001033136.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P333749 | BC055754.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P333749 | AK151090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P333749 | AK153322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P333749 | AK153369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P333749 | NM_001033136.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233027 | BC055754.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233027 | AK151090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233027 | AK153322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233027 | AK153369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233027 | NM_001033136.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233027 | BC055754.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233027 | AK151090.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233027 | AK153322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233027 | AK153369.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233027 | NM_001033136.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144128 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110136 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01820843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21567968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02021296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02021291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02021292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01631513 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10486182 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486173 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486178 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486183 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486175 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486179 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486174 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486184 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486181 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486177 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486176 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452704_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428196_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10486172 | - | - |
| Mouse Genome 430 2.0 Array | 1458722_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428196_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1452704_at | - | - |
| Mouse Genome 430 2.0 Array | 1452704_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428196_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1458722_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67809_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67809_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200036929.mm.1 | NM_001033136.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200036929.mm.1 | NM_001033136.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200036929.mm.1 | TC0200004423.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428196_PM_a_at | - | - |
| Mouse Expression Set 430 | 1428196_a_at | - | - |
| Mouse Expression Set 430 | 1452704_at | - | - |
| Mouse Expression Set 430 | 1452704_PM_at | - | - |
| Murine Genome U74v2 Set | 95078_PM_at | - | - |
| Murine Genome U74v2 Set | 95078_at | - | - |
| Murine Genome U74v2 Set | 169610_r_at | - | - |
| Murine Genome U74v2 Set | 169610_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452704_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428196_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452704_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428196_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452704_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428196_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428196_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452704_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 647267 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2753515 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2753515 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3071764 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3150536 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3071764 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3150536 | - | - |

Rmdn3
Genomic Map