Assay Details
| Gene Symbol: Bmf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138313.3 | NP_612186.2 | 1 - 2 | 141 | 155 | |
| GenBank mRNA | AK084658.1 | - | 1 - 2 | 151 | 155 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bmf
Genomic Map