Assay Details
| Gene Symbol: Dll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019454.3 | NP_062327.2 | 2 - 3 | 699 | 62 | |
| GenBank mRNA | AB043893.1 | - | 2 - 3 | 342 | 62 | |
| AF253469.1 | - | 2 - 3 | 654 | 62 | ||
| AF273454.1 | - | 2 - 3 | 720 | 62 | ||
| AK004739.1 | - | 2 - 3 | 667 | 62 | ||
| AK031699.1 | - | 2 - 3 | 668 | 62 | ||
| AK052322.1 | - | 2 - 3 | 700 | 62 | ||
| AK165517.1 | - | 2 - 3 | 668 | 62 | ||
| AK171717.1 | - | 2 - 3 | 668 | 62 | ||
| BC042497.1 | - | 2 - 3 | 654 | 4017786 | 62 | |
| BC049130.1 | - | 2 - 3 | 659 | 6825525 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dll4
Genomic Map