Assay Details
| Gene Symbol: Dll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019454.3 | NP_062327.2 | 6 - 7 | 1210 | 86 | |
| GenBank mRNA | AB043893.1 | - | 6 - 7 | 853 | 86 | |
| AF253469.1 | - | 6 - 7 | 1165 | 86 | ||
| AF273454.1 | - | 6 - 7 | 1231 | 86 | ||
| AK004739.1 | - | 6 - 7 | 1178 | 86 | ||
| AK031699.1 | - | 6 - 7 | 1179 | 86 | ||
| AK052322.1 | - | 6 - 7 | 1211 | 86 | ||
| AK165517.1 | - | 6 - 7 | 1178 | 86 | ||
| AK171717.1 | - | 6 - 7 | 1177 | 86 | ||
| BC042497.1 | - | 6 - 7 | 1165 | 4017786 | 86 | |
| BC049130.1 | - | 6 - 7 | 1170 | 6825525 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dll4
Genomic Map