Assay Details
| Gene Symbol: Gchfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177157.4 | NP_796131.1 | 2 - 3 | 174 | 62 | |
| GenBank mRNA | AK028116.1 | - | 2 - 3 | 175 | 62 | |
| AK028122.1 | - | 2 - 3 | 175 | 62 | ||
| BC096406.1 | - | 2 - 3 | 162 | 1511037 | 62 | |
| BC132450.1 | - | 2 - 3 | 170 | 40130727 | 62 | |
| BC132478.1 | - | 2 - 3 | 170 | 40130755 | 62 | |
More Information
Set Membership: |
|

Gchfr
Genomic Map