Assay Details
| Gene Symbol: Pla2g4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024145.2 | NP_001019316.1 | 11 - 12 | 1153 | 92 | |
| GenBank mRNA | AB195278.1 | - | 11 - 12 | 1153 | 92 | |
| AK076378.1 | - | 7 - 8 | 561 | 92 | ||
| BC039947.1 | - | 11 - 12 | 1002 | 92 | ||
| BC145426.1 | - | 11 - 12 | 1105 | 9053984 | 92 | |
| BC145427.1 | - | 11 - 12 | 1102 | 92 | ||
| BC145428.1 | - | 11 - 12 | 1105 | 9053986 | 92 | |
| BC150771.1 | - | 11 - 12 | 1105 | 9087682 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g4f
Genomic Map