| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200037405.mm.1 |
NM_001024145.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037405.mm.1 |
NM_001024145.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037405.mm.1 |
TC0200004457.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200037403.mm.1 |
NM_001024145.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037403.mm.1 |
NM_001024145.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037403.mm.1 |
TC0200004457.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200037406.mm.1 |
NM_001024145.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037406.mm.1 |
NM_001024145.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200037406.mm.1 |
TC0200004457.mm.1 |
MTA TC |