Assay Details
| Gene Symbol: Pla2g4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024145.2 | NP_001019316.1 | 2 - 3 | 298 | 62 | |
| GenBank mRNA | AB195278.1 | - | 2 - 3 | 298 | 62 | |
| BC039947.1 | - | 2 - 3 | 147 | 62 | ||
| BC145426.1 | - | 2 - 3 | 250 | 9053984 | 62 | |
| BC145427.1 | - | 2 - 3 | 250 | 62 | ||
| BC145428.1 | - | 2 - 3 | 253 | 9053986 | 62 | |
| BC150771.1 | - | 2 - 3 | 253 | 9087682 | 62 | |
| DQ904008.1 | - | 2 - 3 | 298 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g4f
Genomic Map