Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200008595.mm.1 |
NM_028365.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008595.mm.1 |
NM_028365.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008595.mm.1 |
TC1200001218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008593.mm.1 |
NM_001199785.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008593.mm.1 |
NM_001199785.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008593.mm.1 |
NM_028365.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008593.mm.1 |
NM_028365.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008593.mm.1 |
TC1200001218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008597.mm.1 |
NM_001199785.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008597.mm.1 |
NM_001199785.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008597.mm.1 |
NM_028365.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008597.mm.1 |
NM_028365.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008597.mm.1 |
TC1200001218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008594.mm.1 |
NM_001199785.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008594.mm.1 |
NM_001199785.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008594.mm.1 |
NM_028365.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008594.mm.1 |
NM_028365.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008594.mm.1 |
TC1200001218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008596.mm.1 |
NM_001199785.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008596.mm.1 |
NM_001199785.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008596.mm.1 |
NM_028365.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008596.mm.1 |
NM_028365.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008596.mm.1 |
TC1200001218.mm.1 |
MTA TC |