Assay Details
| Gene Symbol: Edaradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133643.4 | NP_598398.3 | 4 - 5 | 695 | 66 | |
| GenBank mRNA | AF358671.1 | - | 4 - 5 | 689 | 66 | |
| AK041545.1 | - | 4 - 5 | 629 | 66 | ||
| AK042128.1 | - | 4 - 5 | 353 | 66 | ||
| AK042414.1 | - | 4 - 5 | 658 | 66 | ||
| AK075786.1 | - | 4 - 5 | 331 | 66 | ||
| AY071863.1 | - | 4 - 5 | 193 | 66 | ||
| BC030683.1 | - | 4 - 5 | 271 | 1344937 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Edaradd
Genomic Map