Assay Details
| Gene Symbol: Tunar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045047.1 | 2 - 3 | 696 | 146 | ||
| NR_125773.1 | 2 - 3 | 448 | 146 | |||
| GenBank mRNA | AK012713.1 | - | 1 - 2 | 344 | 146 | |
| AK045952.1 | - | 2 - 3 | 450 | 146 | ||
| BC059025.1 | - | 2 - 3 | 696 | 146 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578276 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727131 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498935 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405417 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 769507 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 201546 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749150 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750835 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706573 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467479 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692331 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519934 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438919 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92207 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339092 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66935 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167916 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78588 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184641 | AK045952.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184641 | BC059025.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07736993 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07736943 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02596823 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02596803 | - | - |
| Whole Mouse Genome G4122A | A_52_P166382 | AK045952.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P166382 | BC059025.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P166382 | AK012713.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P290374 | AK045952.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P290374 | BC059025.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117987 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07736993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10289079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10289083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10376117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09855314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09855310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09956521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09839183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09887969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10289076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10111852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09855321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09839185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09855319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09937191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708754 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09917035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10381118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10111853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10213597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09887966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10381121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10213595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10131892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10213596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10131906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09937196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10051656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09855325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09882063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10381137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10213592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10051657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10381125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708751 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1430951_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441087_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430951_at | - | - |
| Mouse Genome 430 2.0 Array | 1441087_at | - | - |
| Mouse Genome 430 2.0 Array | 1441087_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430951_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69952_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69952_at | - | - |
| Mouse Expression Set 430 | 1430951_at | - | - |
| Mouse Expression Set 430 | 1430951_PM_at | - | - |
| Mouse Expression Set 430 | 1441087_PM_at | - | - |
| Mouse Expression Set 430 | 1441087_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441087_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430951_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441087_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430951_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101740537 | - | - |

Tunar
Genomic Map