Assay Details
| Gene Symbol: 1700061F12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038180.1 | 1 - 2 | 135 | 87 | ||
| GenBank mRNA | AK006852.1 | - | 1 - 2 | 136 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
CUGBP, Elav-like family member 2 |
Celf2 |
B230218O03, B230345P09Rik, C88023, CELF-2, CUG-BP2, Cugbp2, D230046B21Rik, Etr-3, Napor, Napor-2, mETR-3 |
-1.8mb | Anti-sense | Chr. 2 - 6539694 - 7396227 on NCBI Build GRCm38 |
| Downstream | predicted gene 13219 | Gm13219 | OTTMUSG00000011007 | 162.0kb | Sense | Chr. 2 - 9352413 - 9467458 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700061F12Rik
Genomic Map