Assay Details
Gene Symbol: Olfr1015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146571.2 | NP_666782.2 | 1 | 931 | 117 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162802 | NM_146571.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162802 | BC119114.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162802 | BC119142.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367321 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172394 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162802 | NM_146571.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162802 | BC119114.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162802 | BC119142.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379264 | NM_146571.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379264 | BC119114.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P379264 | BC119142.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399316 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399318 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399313 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399332 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399342 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399347 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399343 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399329 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399358 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399304 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399340 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399336 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399333 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399354 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399339 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399322 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399297 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399300 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399299 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399301 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399303 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399357 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399302 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399352 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399324 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399335 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399326 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399338 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399356 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399325 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399350 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399327 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172394 | - | - |
Whole Mouse Genome G4122A | A_51_P379264 | NM_146571.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P379264 | BC119114.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P379264 | BC119142.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117295 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172394 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01352203 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02087236 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02087233 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01352205 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02087244 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02087237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399302 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399363 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399360 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399301 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01604263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399322 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399325 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399334 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399338 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399300 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02087254 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399332 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399333 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399327 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399335 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399352 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399343 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399303 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399350 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399368 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399345 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399339 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399356 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399302 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399369 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399324 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399318 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02087248 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399355 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399299 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399304 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399313 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399358 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02130228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399309 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399307 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399362 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399311 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02087240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01621696 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399314 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10473465 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10473466 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10473464 | - | - |
Affymetrix Mouse Clariom S Array | PSR0200009957.mm.1 | NM_146571.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009957.mm.1 | NM_146571.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009957.mm.1 | TC0200001249.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 330644 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6290324 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006290324 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224677 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1224677 | - | - |
mouse-6 v1.1 (Illumina) | scl4941.1.1_206-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6290324 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl4941.1.1_206-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6290324 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1224677 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224677 | - | - |
