Assay Details
| Gene Symbol: Zfp583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033249.3 | NP_001028421.1 | 4 - 5 | 651 | 137 | |
| GenBank mRNA | AK038614.1 | - | 4 - 5 | 432 | 137 | |
| AK133380.1 | - | 4 - 5 | 653 | 137 | ||
| BC056221.1 | - | 2 - 3 | 240 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24955008 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24955007 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24955004 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113905 | BC056221.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113905 | NM_001033249.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113905 | AK133380.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077510 | BC056221.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077510 | AV174083.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077510 | NM_001033249.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195521 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76157 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 887219 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131365 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729876 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411888 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1469 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 171415 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120947 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113905 | BC056221.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113905 | NM_001033249.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113905 | AK133380.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077510 | BC056221.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077510 | AV174083.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077510 | NM_001033249.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24954993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24954986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24954995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24954992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24954990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24954987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24954984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24954997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24954991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24954988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24954985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24954989 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06116773 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01476971 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06116744 | - | - |
| Whole Mouse Genome G4122A | A_52_P482274 | BC056221.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482274 | AV174083.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P482274 | NM_001033249.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06116744 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06116773 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24955004 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24955007 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24955008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06632156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06155526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06155528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06155523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24954933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955010 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559772 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559771 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559770 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559779 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559767 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559774 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559769 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559781 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559776 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559778 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559777 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559768 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559775 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559780 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559773 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439861_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559766 | - | - |
| Mouse Genome 430 2.0 Array | 1439861_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439861_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700020066.mm.1 | TC0700002208.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700020066.mm.1 | NM_001033249.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020066.mm.1 | NM_001033249.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020067.mm.1 | TC0700002208.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700020067.mm.1 | NM_001033249.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020067.mm.1 | NM_001033249.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020068.mm.1 | TC0700002208.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700020068.mm.1 | NM_001033249.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020068.mm.1 | NM_001033249.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020071.mm.1 | TC0700002208.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700020071.mm.1 | NM_001033249.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020071.mm.1 | NM_001033249.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020065.mm.1 | TC0700002208.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700020065.mm.1 | NM_001033249.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700020065.mm.1 | NM_001033249.3 | Refseq |
| Mouse Expression Set 430 | 1439861_PM_at | - | - |
| Mouse Expression Set 430 | 1439861_at | - | - |
| Murine Genome U74v2 Set | 167878_f_at | - | - |
| Murine Genome U74v2 Set | 167878_PM_f_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439861_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439861_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 654348 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228620 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3055220 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3055220 | - | - |

Zfp583
Genomic Map