Assay Details
| Gene Symbol: Fam188a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024185.4 | NP_077147.2 | 13 - 14 | 1325 | 147 | |
| GenBank mRNA | AK002900.1 | - | 13 - 14 | 1276 | 147 | |
| AK009891.1 | - | 13 - 14 | 1304 | 147 | ||
| AK010050.1 | - | 13 - 14 | 1291 | 147 | ||
| AK049869.1 | - | 12 - 13 | 1249 | 147 | ||
| AK193641.1 | - | 2 - 3 | 171 | 147 | ||
| BC002225.1 | - | 13 - 14 | 1307 | 147 | ||
| BC027202.1 | - | 13 - 14 | 1229 | 4217391 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam188a
Genomic Map