Assay Details
| Gene Symbol: Mrpl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031808.2 | NP_001026978.2 | 1 - 2 | 217 | 70 | |
| GenBank mRNA | AK003340.1 | - | 1 - 2 | 132 | 70 | |
| AK011837.1 | - | 1 - 2 | 156 | 70 | ||
| AK012636.1 | - | 1 - 2 | 145 | 70 | ||
| AK032370.1 | - | 1 - 2 | 217 | 70 | ||
| BC034728.1 | - | 1 - 2 | 156 | 2811521 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003340.1, AK011837.1, AK032370.1, BC034728.1, NM_001031808.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21069127 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21069121 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21069122 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21069130 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21069123 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969221 | AK032370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969221 | NM_001031808.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969221 | AK013297.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969221 | NM_026044.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209172 | AK032370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209172 | BC034728.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209172 | AK003340.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209172 | AK011837.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209172 | NM_001031808.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209172 | AK032370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209172 | BC034728.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209172 | AK003340.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209172 | AK011837.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P209172 | NM_001031808.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662277 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 90108 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969221 | AK032370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969221 | NM_001031808.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969221 | AK013297.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969221 | NM_026044.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209172 | AK032370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209172 | BC034728.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209172 | AK003340.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209172 | AK011837.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209172 | NM_001031808.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209172 | AK032370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209172 | BC034728.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209172 | AK003340.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209172 | AK011837.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P209172 | NM_001031808.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338377 | AK032370.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338377 | NM_001031808.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338377 | AK013297.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P338377 | NM_026044.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P209172 | AK032370.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P209172 | BC034728.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P209172 | AK003340.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P209172 | AK011837.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P209172 | NM_001031808.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P209172 | AK032370.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P209172 | BC034728.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P209172 | AK003340.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P209172 | AK011837.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P209172 | NM_001031808.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21069131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21069108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21069109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21069111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21069122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21069123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21069113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21069132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21069115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21069133 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00324159 | - | - |
| Whole Mouse Genome G4122A | A_51_P338377 | AK032370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338377 | NM_001031808.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338377 | AK013297.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338377 | NM_026044.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209172 | AK032370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209172 | BC034728.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209172 | AK003340.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209172 | AK011837.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209172 | NM_001031808.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209172 | AK032370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209172 | BC034728.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209172 | AK003340.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209172 | AK011837.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209172 | NM_001031808.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135682 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01545952 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01545930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01545933 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01545939 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01545948 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21069123 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21069122 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21069127 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21069121 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21069130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21069113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01545954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02260302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01545930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21069110 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10480580 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10469866 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480581 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10561141 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10480582 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428590_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428589_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428588_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10561140 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10469856 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10480579 | - | - |
| Mouse Genome 430 2.0 Array | 1428588_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1428589_at | - | - |
| Mouse Genome 430 2.0 Array | 1428590_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428589_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428588_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1428590_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107733_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107733_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200025652.mm.1 | NM_001031808.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025652.mm.1 | NM_001031808.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025652.mm.1 | TC0200003123.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200025651.mm.1 | NM_001031808.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025651.mm.1 | NM_001031808.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025651.mm.1 | TC0200003123.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200025653.mm.1 | NM_001031808.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025653.mm.1 | NM_001031808.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025653.mm.1 | TC0200003123.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200025654.mm.1 | NM_001031808.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025654.mm.1 | NM_001031808.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200025654.mm.1 | TC0200003123.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428590_at | - | - |
| Mouse Expression Set 430 | 1428590_PM_at | - | - |
| Mouse Expression Set 430 | 1428589_PM_at | - | - |
| Mouse Expression Set 430 | 1428588_a_at | - | - |
| Mouse Expression Set 430 | 1428588_PM_a_at | - | - |
| Mouse Expression Set 430 | 1428589_at | - | - |
| Murine Genome U74v2 Set | 99185_at | - | - |
| Murine Genome U74v2 Set | 99185_PM_at | - | - |
| Murine Genome U74v2 Set | 170150_PM_r_at | - | - |
| Murine Genome U74v2 Set | 131319_PM_at | - | - |
| Murine Genome U74v2 Set | 131319_at | - | - |
| Murine Genome U74v2 Set | 170150_r_at | - | - |
| Murine Genome U74v2 Set | 134476_at | - | - |
| Murine Genome U74v2 Set | 134476_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428589_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428590_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428588_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428589_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428588_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428590_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428589_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428588_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428588_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428589_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428590_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428590_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 306837 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221636 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221636 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2424475 | - | - |

Mrpl41
Genomic Map