Assay Details
| Gene Symbol: Myoz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF480442.1 | - | 5 - 6 | 510 | 67 | |
| BC104046.2 | - | 5 - 6 | 432 | 40040057 | 67 | |
| BC104047.1 | - | 5 - 6 | 432 | 40040058 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myoz3
Genomic Map