Assay Details
| Gene Symbol: Olfr1245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146788.2 | NP_666999.2 | 1 | 494 | 158 | |
| GenBank mRNA | BC138384.1 | - | 1 | 494 | 8861404 | 158 |
| BC138385.1 | - | 1 | 494 | 8861405 | 158 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr1245
Genomic Map