Assay Details
| Gene Symbol: Slc28a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022317.3 | NP_071712.3 | 6 - 7 | 784 | 95 | |
| GenBank mRNA | AF305211.1 | - | 6 - 7 | 717 | 95 | |
| AK052784.1 | - | 6 - 7 | 785 | 95 | ||
| AK145077.1 | - | 6 - 7 | 785 | 95 | ||
| BC010472.1 | - | 6 - 7 | 772 | 3586005 | 95 | |
| BC013783.1 | - | 6 - 7 | 775 | 3600786 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc28a3
Genomic Map